Search
Search

Rad17| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044371.2 | NP_001037836.1 | 11 - 12 | 1394 | 111 | |
| NM_001283011.1 | NP_001269940.1 | 12 - 13 | 1599 | 111 | ||
| NM_011233.3 | NP_035363.2 | 11 - 12 | 1522 | 111 | ||
| GenBank mRNA | AF085737.1 | - | 11 - 12 | 1445 | 111 | |
| AJ011923.1 | - | 10 - 11 | 1295 | 111 | ||
| AK134277.1 | - | 11 - 12 | 1349 | 111 | ||
| AK162016.1 | - | 11 - 12 | 1382 | 111 | ||
| AK167358.1 | - | 12 - 13 | 1542 | 111 | ||
| BC050267.1 | - | 8 - 9 | 1034 | 111 | ||
| BC066855.1 | - | 11 - 12 | 1512 | 30473984 | 111 | |
Set Membership: |
|