Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100003851.mm.1 |
NM_001110148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003851.mm.1 |
NM_001110148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003851.mm.1 |
NM_010794.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003851.mm.1 |
NM_010794.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003851.mm.1 |
TC1100000500.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003855.mm.1 |
NM_001110148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003855.mm.1 |
NM_001110148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003855.mm.1 |
NM_010794.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003855.mm.1 |
NM_010794.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003855.mm.1 |
TC1100000500.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003857.mm.1 |
NM_001110148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003857.mm.1 |
NM_001110148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003857.mm.1 |
NM_010794.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003857.mm.1 |
NM_010794.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003857.mm.1 |
TC1100000500.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003854.mm.1 |
NM_001110148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003854.mm.1 |
NM_001110148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003854.mm.1 |
NM_010794.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003854.mm.1 |
NM_010794.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003854.mm.1 |
TC1100000500.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003856.mm.1 |
NM_001110148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003856.mm.1 |
NM_001110148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003856.mm.1 |
NM_010794.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003856.mm.1 |
NM_010794.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003856.mm.1 |
TC1100000500.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003850.mm.1 |
NM_001110148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003850.mm.1 |
NM_001110148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003850.mm.1 |
NM_010794.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003850.mm.1 |
NM_010794.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003850.mm.1 |
TC1100000500.mm.1 |
MTA TC |