Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290741.1 | NP_001277670.1 | 3 - 4 | 386 | 101 | |
NM_212484.2 | NP_997649.1 | 4 - 5 | 755 | 101 | ||
GenBank mRNA | AK181033.1 | - | 2 - 3 | 116 | 101 | |
AK217278.1 | - | 2 - 3 | 116 | 101 | ||
AY043269.1 | - | 3 - 4 | 384 | 101 | ||
BC062950.1 | - | 4 - 5 | 743 | 6840162 | 101 | |
CF746424.1 | - | 2 - 3 | 220 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |