Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007799.3 | NP_031825.2 | 2 - 3 | 434 | 62 | |
GenBank mRNA | AK143581.1 | - | 1 - 2 | 136 | 62 | |
AK145875.1 | - | 2 - 3 | 291 | 62 | ||
AK157907.1 | - | 1 - 2 | 156 | 62 | ||
AK165242.1 | - | 1 - 2 | 156 | 62 | ||
AK165271.1 | - | 1 - 2 | 102 | 62 | ||
BC005432.1 | - | 2 - 3 | 434 | 3601519 | 62 | |
CT010252.1 | - | 1 - 2 | 70 | 62 | ||
X97399.1 | - | 1 - 2 | 121 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |