Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081125.1 | NP_001074594.1 | 7 - 8 | 1396 | 80 | |
GenBank mRNA | AM261668.1 | - | 4 - 5 | 621 | 80 | |
AM261669.1 | - | 4 - 5 | 640 | 80 | ||
AM261670.1 | - | 5 - 6 | 727 | 80 | ||
AM262741.1 | - | 5 - 6 | 678 | 80 | ||
AM262742.1 | - | 6 - 7 | 913 | 80 | ||
AM262743.1 | - | 6 - 7 | 1010 | 80 | ||
AM262744.1 | - | 8 - 9 | 1227 | 80 | ||
BC082604.1 | - | 7 - 8 | 1337 | 30672479 | 80 | |
BC085190.1 | - | 7 - 8 | 1312 | 30694633 | 80 | |
X99104.1 | - | 7 - 8 | 1156 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |