Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009227.3 | NP_033253.1 | 3 - 4 | 208 | 131 | |
GenBank mRNA | AK019462.1 | - | 3 - 4 | 192 | 131 | |
AW492281.1 | - | 2 - 3 | 131 | |||
BC008262.1 | - | 3 - 4 | 184 | 2650413 | 131 | |
BC051207.1 | - | 3 - 4 | 166 | 1396204 | 131 | |
BC055765.1 | - | 3 - 4 | 150 | 6407944 | 131 | |
BY168551.1 | - | 3 - 4 | 208 | 131 | ||
X65704.1 | - | 3 - 4 | 112 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |