Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alppl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007433.3 | NP_031459.3 | 2 - 3 | 344 | 71 | |
GenBank mRNA | AK145571.1 | - | 3 - 4 | 253 | 71 | |
AY054302.1 | - | 2 - 3 | 185 | 71 | ||
BC052874.1 | - | 2 - 3 | 339 | 30033693 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |