Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172644.3 | NP_766232.1 | 15 - 16 | 2186 | 68 | |
GenBank mRNA | AK030952.1 | - | 15 - 16 | 2175 | 68 | |
AK033634.1 | - | 13 - 14 | 3197 | 68 | ||
AK037337.1 | - | 15 - 16 | 2205 | 68 | ||
AK083300.1 | - | 4 - 5 | 595 | 68 | ||
AK138809.1 | - | 15 - 16 | 2187 | 68 | ||
AK154133.1 | - | 15 - 16 | 2214 | 68 | ||
AK154348.1 | - | 15 - 16 | 2215 | 68 | ||
AK164885.1 | - | 15 - 16 | 2157 | 68 | ||
BC046429.1 | - | 8 - 9 | 951 | 68 | ||
BC057943.1 | - | 13 - 14 | 2335 | 68 | ||
BC076606.1 | - | 15 - 16 | 2178 | 30625515 | 68 | |
BC089191.1 | - | 15 - 16 | 2160 | 30473852 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |