Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146062.3 | NP_666174.2 | 3 - 4 | 412 | 72 | |
GenBank mRNA | BC031486.1 | - | 3 - 4 | 365 | 3157250 | 72 |
BG145871.1 | - | 3 - 4 | 334 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |