Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025937.4 | NP_080213.3 | 1 - 2 | 614 | 68 | |
GenBank mRNA | AK011492.1 | - | 1 - 2 | 565 | 68 | |
AK012826.1 | - | 1 - 2 | 547 | 68 | ||
AK038344.1 | - | 1 - 2 | 562 | 68 | ||
AK076125.1 | - | 1 - 2 | 550 | 68 | ||
AK089943.1 | - | 1 - 2 | 542 | 68 | ||
AY388959.1 | - | 1 - 2 | 565 | 68 | ||
BC026774.1 | - | 1 - 2 | 582 | 4019392 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |