Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139311.2 | NP_647472.2 | 7 - 8 | 928 | 77 | |
GenBank mRNA | AK143477.1 | - | 3 - 4 | 371 | 77 | |
AY050217.1 | - | 7 - 8 | 1075 | 77 | ||
BY731745.1 | - | 3 - 4 | 367 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |