Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159598.1 | NP_001153070.1 | 6 - 7 | 2209 | 79 | |
NM_009733.2 | NP_033863.2 | 6 - 7 | 2209 | 79 | ||
GenBank mRNA | AF009011.1 | - | 6 - 7 | 79 | ||
BC011544.1 | - | 6 - 7 | 1928 | 79 | ||
BC055491.1 | - | 2 - 3 | 596 | 79 | ||
BC113171.1 | - | 6 - 7 | 1941 | 40061055 | 79 | |
BG803725.1 | - | 1 - 2 | 240 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |