Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159598.1 | NP_001153070.1 | 7 - 8 | 2379 | 69 | |
NM_009733.2 | NP_033863.2 | 7 - 8 | 2379 | 69 | ||
GenBank mRNA | AF009011.1 | - | 7 - 8 | 2331 | 69 | |
BC011544.1 | - | 7 - 8 | 2098 | 69 | ||
BC055491.1 | - | 3 - 4 | 766 | 69 | ||
BC113171.1 | - | 7 - 8 | 2111 | 40061055 | 69 | |
BG803725.1 | - | 2 - 3 | 410 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |