Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029447.2 | NP_083723.1 | 8 - 9 | 1486 | 57 | |
GenBank mRNA | AK079090.1 | - | 8 - 9 | 1487 | 57 | |
AK138399.1 | - | 8 - 9 | 1459 | 57 | ||
AK146364.1 | - | 8 - 9 | 1435 | 57 | ||
AK170508.1 | - | 8 - 9 | 1487 | 57 | ||
AK171432.1 | - | 8 - 9 | 1487 | 57 | ||
BC016224.1 | - | 8 - 9 | 1459 | 4486941 | 57 | |
BC025520.1 | - | 1 - 2 | 345 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |