Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010668.2 | NP_034798.2 | 2 - 3 | 933 | 62 | |
GenBank mRNA | AK132476.1 | - | 2 - 3 | 935 | 62 | |
AK161078.1 | - | 2 - 3 | 932 | 62 | ||
AK161098.1 | - | 2 - 3 | 935 | 62 | ||
BC120485.1 | - | 2 - 3 | 899 | 40129408 | 62 | |
BC139485.1 | - | 2 - 3 | 899 | 8860587 | 62 | |
X74784.1 | - | 2 - 3 | 899 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |