Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700003487.mm.1 |
NR_037906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003487.mm.1 |
NR_037906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003487.mm.1 |
TC1700000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003489.mm.1 |
NR_037906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003489.mm.1 |
NR_037906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003489.mm.1 |
TC1700000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003490.mm.1 |
NR_037906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003490.mm.1 |
NR_037906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003490.mm.1 |
TC1700000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003488.mm.1 |
NR_037906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003488.mm.1 |
NR_037906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003488.mm.1 |
TC1700000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003486.mm.1 |
NR_037905.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003486.mm.1 |
NR_037905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003486.mm.1 |
NR_037906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003486.mm.1 |
NR_037906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003486.mm.1 |
TC1700000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NM_025881.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NM_025881.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NM_028190.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NM_028190.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NR_037905.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NR_037905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NR_037906.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
NR_037906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003494.mm.1 |
TC1700000410.mm.1 |
MTA TC |