Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033074.3 | NP_149065.2 | 13 - 14 | 1660 | 153 | |
GenBank mRNA | AK010256.1 | - | 13 - 14 | 1627 | 153 | |
AK011146.2 | - | 13 - 14 | 1661 | 153 | ||
AK031064.1 | - | 13 - 14 | 1624 | 153 | ||
AK152945.1 | - | 13 - 14 | 1621 | 153 | ||
AK153314.1 | - | 13 - 14 | 1632 | 153 | ||
AK166693.1 | - | 13 - 14 | 1627 | 153 | ||
AK167597.1 | - | 13 - 14 | 1669 | 153 | ||
AK168969.1 | - | 13 - 14 | 1620 | 153 | ||
BC004621.1 | - | 6 - 7 | 747 | 153 | ||
BC055371.1 | - | 13 - 14 | 1646 | 30060749 | 153 | |
BC060176.1 | - | 1755 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |