Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU604548.1 | - | 11 - 12 | 827 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |