Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198927.3 | NP_945121.1 | 7 - 8 | 834 | 73 | |
GenBank mRNA | AK017450.1 | - | 7 - 8 | 834 | 73 | |
AK132579.1 | - | 7 - 8 | 836 | 73 | ||
AY459348.1 | - | 7 - 8 | 835 | 73 | ||
BC049243.1 | - | 2 - 3 | 199 | 73 | ||
BC138091.1 | - | 10 - 11 | 1195 | 8861110 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |