Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076365.1 | - | 9 - 10 | 2185 | 100 | |
AK139088.1 | - | 11 - 12 | 1475 | 100 | ||
U81490.1 | - | 11 - 12 | 1209 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |