Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013891.4 | NP_038919.1 | 3 - 4 | 1031 | 54 | |
GenBank mRNA | AB019436.1 | - | 3 - 4 | 967 | 54 | |
AK078863.1 | - | 3 - 4 | 1032 | 54 | ||
BC012648.1 | - | 3 - 4 | 1032 | 4218741 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |