Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034885.3 | NP_001030057.1 | 1 - 2 | 660 | 77 | |
GenBank mRNA | AK132521.1 | - | 1 - 2 | 661 | 77 | |
BC139043.1 | - | 1 - 2 | 367 | 8862065 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |