Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016704.2 | NP_057913.2 | 9 - 10 | 1422 | 115 | |
GenBank mRNA | AF184900.1 | - | 9 - 10 | 1423 | 115 | |
BC011251.1 | - | 9 - 10 | 1411 | 4196330 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |