Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010601.3 | NP_034731.3 | 9 - 10 | 1702 | 69 | |
GenBank mRNA | AY380579.1 | - | 9 - 10 | 1702 | 69 | |
BC141013.1 | - | 9 - 10 | 1817 | 9055625 | 69 | |
BC145145.1 | - | 9 - 10 | 1817 | 9053696 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |