Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253768.1 | NP_001240697.1 | 8 - 9 | 1287 | 83 | |
NM_001253769.1 | NP_001240698.1 | 7 - 8 | 1183 | 83 | ||
NM_001253770.1 | NP_001240699.1 | 7 - 8 | 1032 | 83 | ||
NM_138744.3 | NP_620083.1 | 7 - 8 | 1032 | 83 | ||
GenBank mRNA | AF532969.1 | - | 6 - 7 | 827 | 83 | |
AK031356.1 | - | 8 - 9 | 1011 | 83 | ||
AK043865.1 | - | 7 - 8 | 1136 | 83 | ||
AK049080.1 | - | 7 - 8 | 1184 | 83 | ||
AK077112.1 | - | 8 - 9 | 1072 | 83 | ||
AK173064.1 | - | 7 - 8 | 1019 | 83 | ||
BC021749.1 | - | 7 - 8 | 987 | 4165430 | 83 | |
BC031527.1 | - | 7 - 8 | 995 | 4008624 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |