Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172742.2 | NP_766330.2 | 8 - 9 | 948 | 82 | |
GenBank mRNA | BC041672.1 | - | 8 - 9 | 920 | 82 | |
BC055074.1 | - | 6 - 7 | 612 | 30052208 | 82 | |
BC058196.1 | - | 1 - 2 | 50 | 82 | ||
CF734341.1 | - | 3 - 4 | 327 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |