Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033439.3 | NP_001028611.2 | 11 - 12 | 1574 | 97 | |
NM_001252132.1 | NP_001239061.1 | 11 - 12 | 1574 | 97 | ||
GenBank mRNA | AK076431.1 | - | 2 - 3 | 173 | 97 | |
AK129264.1 | - | 11 - 12 | 1563 | 97 | ||
AK136026.1 | - | 11 - 12 | 1180 | 97 | ||
AK143909.1 | - | 11 - 12 | 1573 | 97 | ||
AK160902.1 | - | 9 - 10 | 950 | 97 | ||
AK162105.1 | - | 1 - 2 | 81 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |