Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028039.2 | NP_082315.3 | 1 - 2 | 161 | 66 | |
GenBank mRNA | AK050845.1 | - | 1 - 2 | 112 | 66 | |
AK144685.1 | - | 1 - 2 | 117 | 66 | ||
BC033303.1 | - | 1 - 2 | 76 | 3675568 | 66 | |
BC071275.1 | - | 1 - 2 | 94 | 6331551 | 66 | |
CF168369.1 | - | 1 - 2 | 144 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |