Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Negr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039094.3 | NP_001034183.1 | 5 - 6 | 1172 | 93 | |
NM_177274.4 | NP_796248.1 | 5 - 6 | 1172 | 93 | ||
GenBank mRNA | AJ487032.1 | - | 5 - 6 | 792 | 93 | |
AK030503.1 | - | 5 - 6 | 998 | 93 | ||
AK035218.1 | - | 5 - 6 | 1008 | 93 | ||
AK122576.2 | - | 4 - 5 | 515 | 93 | ||
AK142298.1 | - | 5 - 6 | 866 | 93 | ||
AK147253.1 | - | 5 - 6 | 1173 | 93 | ||
BC146005.1 | - | 5 - 6 | 957 | 40131308 | 93 | |
BC146007.1 | - | 5 - 6 | 956 | 40131310 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |