Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177765.3 | NP_808433.2 | 12 - 13 | 2022 | 71 | |
GenBank mRNA | AK029565.1 | - | 12 - 13 | 2021 | 71 | |
AK161519.1 | - | 12 - 13 | 1845 | 71 | ||
AM690757.1 | - | 11 - 12 | 1723 | 71 | ||
BC145749.1 | - | 11 - 12 | 1630 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |