Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700029577.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029577.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029577.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029582.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029582.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029582.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029583.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029583.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029583.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029584.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029584.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029584.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029581.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029581.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029581.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029576.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029576.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029576.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029587.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029587.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029587.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029579.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029579.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029579.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029580.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029580.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029580.mm.1 |
TC0700003485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029586.mm.1 |
NM_001024703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029586.mm.1 |
NM_001024703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029586.mm.1 |
TC0700003485.mm.1 |
MTA TC |