Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_001253768.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_001253768.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_001253769.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_001253769.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_001253770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_001253770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_138744.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
NM_138744.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012075.mm.1 |
TC0300001491.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_001253768.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_001253768.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_001253769.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_001253769.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_001253770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_001253770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_138744.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
NM_138744.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300012076.mm.1 |
TC0300001491.mm.1 |
MTA TC |