Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253769.1 | NP_001240698.1 | 1 - 2 | 295 | 74 | |
GenBank mRNA | AK043865.1 | - | 1 - 2 | 248 | 74 | |
AK049080.1 | - | 1 - 2 | 296 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |