Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008486.2 | NP_032512.2 | 2 - 3 | 861 | 77 | |
GenBank mRNA | AK143746.1 | - | 2 - 3 | 804 | 77 | |
BC005431.1 | - | 2 - 3 | 827 | 3593307 | 77 | |
BC017011.1 | - | 2 - 3 | 885 | 3590117 | 77 | |
BC040792.1 | - | 2 - 3 | 885 | 3590117 | 77 | |
U77083.1 | - | 2 - 3 | 793 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |