Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030131.3 | NP_084407.1 | 2 - 3 | 201 | 61 | |
GenBank mRNA | AK021301.1 | - | 2 - 3 | 155 | 61 | |
AK144157.1 | - | 2 - 3 | 162 | 61 | ||
AK165179.1 | - | 2 - 3 | 147 | 61 | ||
AK166153.1 | - | 2 - 3 | 145 | 61 | ||
BC040509.1 | - | 2 - 3 | 141 | 61 | ||
BC065395.1 | - | 2 - 3 | 145 | 876056 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |