Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010374.3 | NP_034504.1 | 3 - 4 | 389 | 74 | |
GenBank mRNA | BC100311.1 | - | 3 - 4 | 389 | 30523018 | 74 |
BC145435.1 | - | 3 - 4 | 365 | 9053993 | 74 | |
J03257.1 | - | 3 - 4 | 399 | 74 | ||
X12823.1 | - | 3 - 4 | 340 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |