Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019670.1 | NP_062644.1 | 1 - 2 | 115 | 77 | |
GenBank mRNA | AF094519.1 | - | 1 - 2 | 115 | 77 | |
AK138694.1 | - | 1 - 2 | 145 | 77 | ||
AK166225.1 | - | 1 - 2 | 145 | 77 | ||
AK166882.1 | - | 1 - 2 | 171 | 77 | ||
AK220493.1 | - | 1 - 2 | 179 | 77 | ||
BC085191.1 | - | 1 - 2 | 225 | 30693873 | 77 | |
BC086779.1 | - | 1 - 2 | 190 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |