Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008491.1 | NP_032517.1 | 5 - 6 | 608 | 63 | |
GenBank mRNA | AK002932.1 | - | 5 - 6 | 641 | 63 | |
AK149774.1 | - | 5 - 6 | 640 | 63 | ||
BC020275.1 | - | 5 - 6 | 608 | 63 | ||
BC132069.1 | - | 5 - 6 | 608 | 40130346 | 63 | |
BC132071.1 | - | 5 - 6 | 608 | 40130348 | 63 | |
X14607.1 | - | 5 - 6 | 608 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |