Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123362.1 | NP_001116834.1 | 1 - 2 | 283 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Mus musculus Inventoried Probe spans exons |