Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003685.3 | NP_001003685.2 | 9 - 10 | 974 | 102 | |
GenBank mRNA | BC120748.1 | - | 9 - 10 | 904 | 40129671 | 102 |
BC120774.1 | - | 9 - 10 | 904 | 40129697 | 102 | |
BC144892.1 | - | 9 - 10 | 904 | 102 | ||
L07379.1 | - | 9 - 10 | 907 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |