Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207298.2 | NP_997181.1 | 5 - 6 | 804 | 91 | |
GenBank mRNA | AK155384.1 | - | 5 - 6 | 800 | 91 | |
BC052176.1 | - | 5 - 6 | 765 | 91 | ||
BC138847.1 | - | 5 - 6 | 783 | 8861869 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |