Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080944.3 | NP_001074413.2 | 22 - 23 | 2588 | 81 | |
GenBank mRNA | AK155448.1 | - | 22 - 23 | 2598 | 81 | |
AK169967.1 | - | 22 - 23 | 2616 | 81 | ||
BC130260.1 | - | 4 - 5 | 562 | 81 | ||
BC152337.1 | - | 4 - 5 | 562 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |