Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080944.3 | NP_001074413.2 | 6 - 7 | 467 | 117 | |
GenBank mRNA | AK040903.1 | - | 7 - 8 | 636 | 117 | |
AK086607.1 | - | 6 - 7 | 501 | 117 | ||
AK155448.1 | - | 6 - 7 | 478 | 117 | ||
AK169967.1 | - | 6 - 7 | 490 | 117 | ||
AK173307.1 | - | 6 - 7 | 467 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |