Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010190.1 | NP_034320.1 | 2 - 3 | 200 | 76 | |
GenBank mRNA | AK010913.1 | - | 1 - 2 | 198 | 76 | |
AK151676.1 | - | 2 - 3 | 208 | 76 | ||
BC107220.1 | - | 2 - 3 | 200 | 40051808 | 76 | |
BC107221.1 | - | 2 - 3 | 200 | 40051810 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |