Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040682.1 | NP_001035772.1 | 4 - 5 | 450 | 71 | |
NM_053155.2 | NP_444385.2 | 4 - 5 | 450 | 71 | ||
NR_104435.1 | 4 - 5 | 450 | 71 | |||
NR_104436.1 | 4 - 5 | 450 | 71 | |||
NR_104437.1 | 4 - 5 | 450 | 71 | |||
NR_104438.1 | 4 - 5 | 450 | 71 | |||
NR_104439.1 | 4 - 5 | 450 | 71 | |||
GenBank mRNA | AB047978.1 | - | 4 - 5 | 443 | 71 | |
AB059643.1 | - | 4 - 5 | 443 | 71 | ||
AB059644.1 | - | 4 - 5 | 443 | 71 | ||
AB059645.1 | - | 4 - 5 | 443 | 71 | ||
AB059646.1 | - | 4 - 5 | 443 | 71 | ||
AB059647.1 | - | 4 - 5 | 443 | 71 | ||
AB059648.1 | - | 4 - 5 | 443 | 71 | ||
AK147777.1 | - | 4 - 5 | 347 | 71 | ||
BC019176.1 | - | 4 - 5 | 381 | 71 | ||
BC096470.1 | - | 4 - 5 | 339 | 71 | ||
BC141280.1 | - | 4 - 5 | 383 | 9055892 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |