Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104436.1 | 12 - 13 | 3019 | 70 | ||
NR_104438.1 | 12 - 13 | 3019 | 70 | |||
NR_104439.1 | 11 - 12 | 2938 | 70 | |||
GenBank mRNA | AB059643.1 | - | 11 - 12 | 2931 | 70 | |
AB059646.1 | - | 12 - 13 | 3012 | 70 | ||
AB059648.1 | - | 12 - 13 | 3012 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |