Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029716.3 | NP_083992.1 | 1 - 2 | 218 | 67 | |
GenBank mRNA | AK007182.1 | - | 1 - 2 | 218 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |