Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271879.1 | NP_001258808.1 | 11 - 12 | 1318 | 62 | |
NM_177694.6 | NP_808362.2 | 12 - 13 | 1360 | 62 | ||
NR_073508.1 | 12 - 13 | 1353 | 62 | |||
GenBank mRNA | AB125740.1 | - | 12 - 13 | 1360 | 62 | |
AB206762.1 | - | 12 - 13 | 1360 | 62 | ||
AK034197.1 | - | 12 - 13 | 1342 | 62 | ||
BC109163.2 | - | 11 - 12 | 1240 | 40047987 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |