Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016716.4 | NP_057925.1 | 7 - 8 | 1145 | 89 | |
GenBank mRNA | AF129738.1 | - | 7 - 8 | 1035 | 89 | |
AK129174.1 | - | 8 - 9 | 1533 | 89 | ||
BC027304.1 | - | 7 - 8 | 1170 | 3981170 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |