Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011706.2 | NP_035836.2 | 1 - 2 | 117 | 77 | |
GenBank mRNA | AB021665.1 | - | 1 - 2 | 201 | 77 | |
BB631765.1 | - | 1 - 2 | 117 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mus musculus Probe spans exons |